3-Fluorphenol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A13598.09
A13598.18
A13598.09EA
622
DKK
A13598.09
A13598.18
3-Fluorphenol ≥98%
3-Fluorphenol
Purity: 98%
CAS No.: 372-20-3
Molecular Formula: C6H5FO
Formula Weight: 112.10
Flash Point: 71°(159°F)
Refractive Index: 1.5140
Odour: Phenol-like
Melting Point: 12-14°
Boiling Point: 177-179°
Beilstein Reference: 1904536
UN No.: 2922
MDL No.: MFCD00002254
Formel:
C₆H₅FO Molekylvægt: 112.1 g/mol Kogepunkt: 177…179 °C Smeltepunkt: 12…14 °C Massefylde: 1.246 Flammepunkt: 71 °C (159 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002254 CAS-nummer: 372-20-3 EINECS: 206-748-6 UN nummer: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...