2-Fluoranilin 99+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
119270250
119271000
119275000
ACRO119270250EA
306
DKK
ACRO119270250
ACRO119271000
ACRO119275000
2-Fluoranilin 99+%
2-Fluoranilin
Formel:
C₆H₆FN Molekylvægt: 111.12 g/mol Kogepunkt: 182…183 °C (760 mmHg) Smeltepunkt: –29 °C Massefylde: 1.151 g/cm³ Flammepunkt: 60 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007642 CAS-nummer: 348-54-9 EINECS: 206-478-9 UN nummer: 1992 ADR: 3 III |
Specifikationer Testresultater
Appearance | Clear colorless to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5410 to 1.5430 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...