5-Fluoranthranilsyre 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

308940010 308940050
ACRO308940010EA 470 DKK
ACRO308940010 ACRO308940050
5-Fluoranthranilsyre 97%
5-Fluoranthranilsyre
Formel: C₇H₆FNO₂
Kogepunkt: 305 °C (1013 hPa)
Smeltepunkt: 181…185 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00055566
CAS-nummer: 446-08-2
EINECS: 207-159-7

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 181 to 185 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR