3,4-Difluorbenzonitril ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 64248-62-0
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Flash Point: 69°(156°F)
Melting Point: 48-52°
Boiling Point: 180°
Beilstein Reference: 2082224
UN No.: 3439
MDL No.: MFCD00011666
Formel:
C₇H₃F₂N Kogepunkt: 180 °C Smeltepunkt: 48…52 °C Flammepunkt: 69 °C (156 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011666 CAS-nummer: 64248-62-0 EINECS: 264-751-8 UN nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...