5-Fluor-2-nitrobenzaldehyd ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L19327.03 L19327.06
L19327.03EA 894 DKK
L19327.03 L19327.06
5-Fluor-2-nitrobenzaldehyd ≥98%
5-Fluor-2-nitrobenzaldehyd

Purity: 98% CAS No.: 395-81-3 Molecular Formula: C7H4FNO3 Formula Weight: 169.11 Melting Point: 93-96° Boiling Point: 153°/23mm MDL No.: MFCD00153175

Formel: C₇H₄FNO₃
Molekylvægt: 169.11 g/mol
Kogepunkt: 153 °C (23 mmHg)
Smeltepunkt: 93…96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00153175
CAS-nummer: 395-81-3
EINECS: 206-903-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR