Pentafluorbenzaldehyd 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A15490.06
A15490.14
A15490.22
A15490.06EA
678
DKK
A15490.06
A15490.14
A15490.22
Pentafluorbenzaldehyd 98%
Pentafluorobenzaldehyde
Purity: 98+%
CAS No.: 653-37-2
Molecular Formula: C7HF5O
Formula Weight: 196.07
Flash Point: 77°(170°F)
Refractive Index: 1.4510
Melting Point: 24-27°
Boiling Point: 164-166°
Beilstein Reference: 1876550
UN No.: 2811
MDL No.: MFCD00003303
Formel:
C₇HF₅O Kogepunkt: 164…166 °C Smeltepunkt: 24…27 °C Massefylde: 1,630 Flammepunkt: 77 °C (170 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003303 CAS-nummer: 653-37-2 EINECS: 211-502-6 UN nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...