4-Fluorbenzaldehyd ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A15383.14 A15383.22 A15383.36
A15383.14EA 420 DKK
A15383.14 A15383.22 A15383.36
4-Fluorbenzaldehyd ≥98%
4-Fluorobenzaldehyde

Purity: 98% CAS No.: 459-57-4 Molecular Formula: C7H5FO Formula Weight: 124.11 Flash Point: 56°(133°F) Refractive Index: 1.5210 Melting Point: -10° Boiling Point: 180-181° Beilstein Reference: 385857 UN No.: 1989 MDL No.: MFCD00003378

Formel: C₇H₅FO
Kogepunkt: 180…181 °C
Smeltepunkt: –10 °C
Massefylde: 1,175
Flammepunkt: 56 °C (133 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003378
CAS-nummer: 459-57-4
EINECS: 207-293-6
UN nummer: 1989
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR