4-Fluorbenzaldehyd 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
156150500
156152500
156150010
ACRO156150500EA
555
DKK
ACRO156150500
ACRO156152500
ACRO156150010
4-Fluorbenzaldehyd 98+%
4-Fluorobenzaldehyde
Formel:
C₇H₅FO Kogepunkt: 181 °C (758 mmHg) Smeltepunkt: –10 °C Massefylde: 1,15 g/cm³ Flammepunkt: 56 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003378 CAS-nummer: 459-57-4 EINECS: 207-293-6 UN nummer: 1989 ADR: 3,III |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5195 to 1.5215 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...