Pentafluorbenzen ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98+%
CAS No.: 363-72-4
Molecular Formula: C6HF5
Formula Weight: 168.06
Flash Point: 13°(55°F)
Refractive Index: 1.3910
Melting Point: -48°
Boiling Point: 84-85°
Beilstein Reference: 1911549
UN No.: 1993
MDL No.: MFCD00000286
Formel:
C₆HF₅ Kogepunkt: 84…85 °C Smeltepunkt: –48 °C Massefylde: 1,518 Flammepunkt: 13 °C (55 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000286 CAS-nummer: 363-72-4 EINECS: 206-658-7 UN nummer: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...