Bottles, narrow neck, square, PP 'Clear Grip', with thread
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
310-86456 310-86457 310-86458 310-86459
215-2879EA 5720 DKK
215-2879 215-2880 215-2881 215-2882
Bottles, narrow neck, square, PP 'Clear Grip', with thread
Flasker Plastflasker
Transparent polypropylene.
  • Low weight
  • Stiffness
  • Impact resistance
  • Easily recyclable
  • Leakproof
  • Suitable for food
  • Halal certified
Good transparency and glaze (ideal for product presentations). Outstanding chemical resistance and thermal stability, balanced stiffness, and impact resistance.