Control seal, close-it tape
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5303-3019 5303-2019 5303-0017 5303-1017
312-0092EA 1780 DKK
312-0092 312-0091 312-0089 312-0090
Control seal, close-it tape
Etiketter
Standard close-it and close-it food & pharma control seals are also available on a continuous roll. This offers the flexibility to provide a small length of tape to seal a small hole or to mark a specific point. Alternatively, longer pieces can be used to repair a long tear quickly, reliably and easily. With its extremely strong adhesive strength, close-it tapes adhere to a range of different surfaces, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
  • Customisable
  • White with tear-off lines
  • Available in the classic close-it design, as well as close-it tape food & pharma, with a food-compatible adhesive
  • Can be labelled with permanent ink providing a gas-proof seal