Bowls
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
074.001 074.002 074.005 074.020 074.035 074.060 074.085 074.125 074.250 074.500
RSGA074.001EA 571 DKK
RSGA074.001 RSGA074.002 RSGA074.005 RSGA074.020 RSGA074.035 RSGA074.060 RSGA074.085 RSGA074.125 RSGA074.250 RSGA074.500
Bowls
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Made of stainless steel.
  • Fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
With flat bottom.