Graduated pipettes, type 2, Class AS, MBL
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PRA810AS PRA812AS PRA814AS PRA816AS PRA818AS
612-3965EA 76.6 DKK
612-3965 612-3966 612-3967 612-3968 612-3969
Graduated pipettes, type 2, Class AS, MBL
Pipetter Pipetter med inddeling
Soda lime glass.
  • Calibrated to deliver from any graduation line down to zero at the jet
  • Graduations in permanent amber stain
DIN EN ISO 835
Certificeringer: DIN EN ISO 835.