Instrument trays, rectangular
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
701.252 701.220 701.242 701.235 701.230 701.228 701.226 701.222 701.217 701.224
RSGA701.252EA 2260 DKK
RSGA701.252 RSGA701.220 RSGA701.242 RSGA701.235 RSGA701.230 RSGA701.228 RSGA701.226 RSGA701.222 RSGA701.217 RSGA701.224
Instrument trays, rectangular
Bakker/Tallerkener/Potter Instrumentbakker/-skåle
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance