Magnetiske omrørerstave, dobbelt ende, PTFE-coatet
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
775 776
KART775EA 184 DKK
KART775 KART776
Magnetiske omrørerstave, dobbelt ende, PTFE-coatet
Omrørere Omrøremagneter
Double ended magnetic stirring bars provide excellent centering, limited contact surface and high turbulence even at low speeds.
  • Suitable for use with foodstuffs
  • Ideal for test tubes or for eccentric stirring movements