Flasker, bred åbning, firkantede, med skruelåg
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0342-0050 0342-0100 0342-2500 0342-1000 0342-0500 0342-0250 0342-4000 0342-1500 0342-0750
215-3209EA 42 DKK
215-3209 215-3210 BURK0342-2500 331-0011 300-0099 BURK0342-0250 BURK0342-4000 BURK0342-1500 BURK0342-0750
Flasker, bred åbning, firkantede, med skruelåg
Flasker Plastflasker
HDPE, transparent.
  • Tamper evident blue screw cap
  • Cap absolutely tight closing
  • UN approval for bulk goods (except bottle with 50 ml capacity)
  • Temperature resistance from −50 °C up to +110 C°
  • Suitable for use with foodstuffs
The reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.