Flasker, bred åbning, med skruelåg, LaboPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0319-1000 0319-0250 0319-0500 0319-0100 0318-0250 0318-0500 0320-0500 0318-1000 0318-0200 0320-1000 0318-0750 0318-0100 0318-0050 0320-0250
300-1018EA 137 DKK
300-1018 331-2612 331-2613 331-2611 300-0118 300-0174 BURK0320-0500 BURK0318-1000 BURK0318-0200 BURK0320-1000 BURK0318-0750 BURK0318-0100 BURK0318-0050 BURK0320-0250
Flasker, bred åbning, med skruelåg, LaboPlast®
Flasker Plastflasker
LDPE, translucent or amber, PP, translucent.
  • Amber coloured for light sensitive substances
  • Leakproof
  • Suitable for use with foodstuffs
Sturdy bottle with wide-necked opening and screw cap. Particularly suitable for viscous media, powder, granules and pastes.