Apparatus - Antonacopoulos
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05434000 05434001 05434002 05434004 05434005
LENZ05434000EA 6200 DKK
LENZ05434000 LENZ05434001 LENZ05434002 LENZ05434004 LENZ05434005
Apparatus - Antonacopoulos
Destillations komponenter Destillationssæt
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes