3-Chloranilin ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A12019.22
A12019.36
A12019.22EA
348
DKK
A12019.22
A12019.36
3-Chloranilin ≥99%
3-Chloranilin
Purity: 99%
CAS No.: 108-42-9
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 123°(253°F)
Refractive Index: 1.5940
Odour: Characteristic
Melting Point: -10°
Boiling Point: 230°
Beilstein Reference: 605969
Merck Reference: 14,2118
UN No.: 2019
MDL No.: MFCD00007765
Formel:
C₆H₆ClN Kogepunkt: 230 °C Smeltepunkt: –10 °C Massefylde: 1,210 Flammepunkt: 123 °C (253 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007765 CAS-nummer: 108-42-9 EINECS: 203-581-0 UN nummer: 2019 ADR: 6.1,II Merck Index: 14,2118 |