2-Chloranilin 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
108570010
108572500
108570050
ACRO108570010EA
733
DKK
ACRO108570010
ACRO108572500
ACRO108570050
2-Chloranilin 98+%
2-Chloranilin
Formel:
C₆H₆ClN Kogepunkt: 209 °C (760 mmHg) Smeltepunkt: –3 °C Massefylde: 1,21 g/cm³ Flammepunkt: 98 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007656 CAS-nummer: 95-51-2 EINECS: 202-426-4 UN nummer: 2019 ADR: 6.1,II Merck Index: 13,02136 |
Specifikationer Testresultater
Appearance | Clear light yellow to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5870 to 1.5890 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...