2-Chlor-1-iodbenzen ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 99%
CAS No.: 615-41-8
Molecular Formula: C6H4ClI
Formula Weight: 238.46
Flash Point: 111°(231°F)
Refractive Index: 1.6345
Melting Point: 1°
Boiling Point: 108-110°/15mm
Beilstein Reference: 1905116
MDL No.: MFCD00001033
Formel:
C₆H₄ClI Kogepunkt: 108…110 °C (15 mmHg) Smeltepunkt: 1 °C Massefylde: 1,954 Flammepunkt: 111 °C (231 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001033 CAS-nummer: 615-41-8 EINECS: 210-423-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...