2-Chlor-3,5-dinitrobenzoesyre, (max. 5% H₂O) 97% (dry weight)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A15675.14 A15675.22
A15675.14EA 272 DKK
A15675.14 A15675.22
2-Chlor-3,5-dinitrobenzoesyre, (max. 5% H₂O) 97% (dry weight)
2-Chlor-3,5-dinitrobenzoesyre

Purity: 97% CAS No.: 2497-91-8 Molecular Formula: C7H3ClN2O6 Formula Weight: 246.56 Melting Point: 196-201° Beilstein Reference: 2146480

Formel: C₇H₃ClN₂O₆
Molekylvægt: 246.56 g/mol
Smeltepunkt: 196…201 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007070
CAS-nummer: 2497-91-8
EINECS: 219-684-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR