4-Chlor-2-nitrobenzoesyre ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A12872.14
A12872.22
A12872.14EA
183
DKK
A12872.14
A12872.22
4-Chlor-2-nitrobenzoesyre ≥97%
4-Chlor-2-nitrobenzoesyre
Purity: 97%
CAS No.: 6280-88-2
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Flash Point:
Formel:
C₇H₄ClNO₄ Smeltepunkt: 142…146 °C Flammepunkt: >100 °C (212 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007214 CAS-nummer: 6280-88-2 EINECS: 228-483-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...