2-Chlor-3-nitrobenzoesyre 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Mesalazine EP Impurity Q

161480050 161480250
ACRO161480050EA 1300 DKK
ACRO161480050 ACRO161480250
2-Chlor-3-nitrobenzoesyre 98%
2-Chlor-3-nitrobenzoesyre
Formel: C₇H₄ClNO₄
Molekylvægt: 201.57 g/mol
Smeltepunkt: 183…188 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007069
CAS-nummer: 3970-35-2
EINECS: 223-590-3

Bestil nu

Specifikationer Testresultater

Appearance White to yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 183 to 188 °C
HPLC ≥97.5 %
GC ≥92 %
Specific optical rotation -31° to -35° (20 °C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR