6-Chloranthranilsyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A10890.14 A10890.06 A10890.09
A10890.14EA 4020 DKK
A10890.14 A10890.06 A10890.09
6-Chloranthranilsyre ≥98%
6-Chloranthranilsyre

Purity: 98% CAS No.: 2148-56-3 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 149-152° Beilstein Reference: 2804041

Formel: C₇H₆ClNO₂
Kogepunkt: 330 °C (1013 hPa)
Smeltepunkt: 149…152 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051530
CAS-nummer: 2148-56-3
EINECS: 218-416-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR