3-Hydroxybenzoesyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
120981000
120985000
120980025
ACRO120981000EA
341
DKK
ACRO120981000
ACRO120985000
ACRO120980025
3-Hydroxybenzoesyre 99%
3-Hydroxybenzoesyre
Formel:
C₇H₆O₃ Kogepunkt: 346 °C (1013 hPa) Smeltepunkt: 201…204 °C Massefylde: 1,473 g/cm³ (20 °C) Flammepunkt: 220 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002506 CAS-nummer: 99-06-9 EINECS: 202-726-5 |
Specifikationer Testresultater
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Melting point | 201 to 205 °C |
HPLC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...