2,3,4-Trimethoxybenzoesyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A14278.09 A14278.18 A14278.30
A14278.09EA 452 DKK
A14278.09 A14278.18 A14278.30
2,3,4-Trimethoxybenzoesyre ≥98%
2,3,4-Trimethoxybenzoesyre

Purity: 98+% CAS No.: 573-11-5 Molecular Formula: C10H12O5 Formula Weight: 212.20 Odour: Odourless Melting Point: 99-102° Beilstein Reference: 2696073 MDL No.: MFCD00002433

Formel: C₁₀H₁₂O₅
Kogepunkt: 110…115 °C (0,04 mmHg)
Smeltepunkt: 99…102 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002433
CAS-nummer: 573-11-5
EINECS: 209-350-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR