GC septa, three layer
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
76001 76002 76006 76007 76008 76009 76010 76013 76015 76016 76020 76022 76003 76004 76005
HAMI76001EA 170 DKK
HAMI76001 HAMI76002 HAMI76006 HAMI76007 HAMI76008 613-1520 HAMI76010 HAMI76013 HAMI76015 HAMI76016 HAMI76020 HAMI76022 HAMI76003 HAMI76004 HAMI76005
GC septa, three layer
Septa
These preconditioned silicone rubber septa are 3,81 mm thick.
  • Average life up to 100 injections with a 26s gauge needle
  • Maximum temperature up to 400 °C