Bowls, conical shape
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4355 4331 4330
USBE4355EA 370 DKK
USBE4355 USBE4331 USBE4330
Bowls, conical shape
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Made of stainless steel.
  • With folded edge; fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose bowl for a wide range of uses.