Svalere, Liebig, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
242516107 242517009 242517103 242517206 242518108 242518202
SCOT242516107EA 520 DKK
SCOT242516107 SCOT242517009 SCOT242517103 SCOT242517206 SCOT242518108 SCOT242518202
Svalere, Liebig, DURAN®
Svaler
DURAN®, borosilicate glass 3.3, clear.
  • Also named as West condenser
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with fused mantle, cone and socket. Water supplied through plastic connectors.