Kolber, rund bund med to halse og standardslib, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
241832604 241832707 241833609 241834408 241835404 241833703 241835601
SCOT241832604EA 453 DKK
SCOT241832604 SCOT241832707 SCOT241833609 SCOT241834408 SCOT241835404 SCOT241833703 SCOT241835601
Kolber, rund bund med to halse og standardslib, DURAN®
Kolber Kolber med rund bund
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one parallel side neck. With matt white labelling area.
*non-DIN ISO size.
Certificeringer: DIN 12394.