Examination gloves, vinyl, Evercare®
Leverandør: Onemed Sverige AB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
295821/6351 295820/6350 295822/6352 295823/6353 295824/6354
113-7909EA 137 DKK
113-7909 113-7908 113-7910 113-7911 113-7912
Examination gloves, vinyl, Evercare®
Handsker
Disposable, powder-free examination gloves for non sterile environments. Ideal for general purposes as well as for food handling (Not suitable for fatty foodstuffs).
  • Non sterile, powder-free
  • Phthalate-free: avoids health concerns
  • Ambidextrous, with rolled rim
  • No latex proteins, good alternative for those suffering from Type I allergies
  • Very soft, good fit, feel and performance
  • In easy-to-use dispenser box of 100 gloves
Colour: Transparent
Length: 240 mm
EN 455; AQL 1,5 Food contact approved
Certificeringer: EN 455; AQL 1,5
Food contact approved.