VWR®, Accessories for Circulators
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
462-0271EA 3530 DKK
462-0271 462-0272 462-0273 462-0274
VWR®, Accessories for Circulators
Cirkulatorsystemer Åbne cirkulatorsystemer
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.