Kolber, rund bund med standardslib, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
241707207 241702706 241702003 241706305 241702509 241702603 241703608 241703702 241704407 241704604 241704707 241705403 241705703 241706408 241705609 241702406 241701307 241701401 241701701 241701804
SCOT241707207EA 1390 DKK
SCOT241707207 SCOT241702706 SCOT241702003 SCOT241706305 SCOT241702509 SCOT241702603 SCOT241703608 SCOT241703702 SCOT241704407 SCOT241704604 SCOT241704707 SCOT241705403 SCOT241705703 SCOT241706408 SCOT241705609 SCOT241702406 SCOT241701307 SCOT241701401 SCOT241701701 SCOT241701804
Kolber, rund bund med standardslib, DURAN®
Kolber Kolber med rund bund
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certificeringer: DIN EN ISO 4797.