Woulff-flasker, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
247104401 247105406 247107304
SCOT247104401EA 1670 DKK
SCOT247104401 189-1110 SCOT247107304
Woulff-flasker, DURAN®
Flasker Woulff-flasker
DURAN®, borosilicate glass 3.3, clear.
  • With bottom tubulature NS 19/26
  • Very good temperature resistance
  • Very good chemical resistance
Bottle with 3 grounded necks.
Certificeringer: DIN 12481.