3-Methoxybenzoesyre 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Anisic acid, 3-Methoxybenzoic acid
152470250
152471000
152475000
ACRO152470250EA
295
DKK
ACRO152470250
ACRO152471000
ACRO152475000
3-Methoxybenzoesyre 98%
3-Methoxybenzoesyre
Formel:
C₈H₈O₃ Molekylvægt: 152.15 g/mol Kogepunkt: 170…172 °C (10 mmHg) Smeltepunkt: 103…108 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002499 CAS-nummer: 586-38-9 EINECS: 209-574-9 |
Specifikationer Testresultater
Appearance | Off-white to yellow to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 103 to 108 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...