2-Methoxybenzoesyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Methoxybenzoic acid

A13513.22 A13513.36
A13513.22EA 518 DKK
A13513.22 A13513.36
2-Methoxybenzoesyre ≥98%
2-Methoxybenzoesyre

Purity: 98+% CAS No.: 579-75-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 99-103° Beilstein Reference: 509929 MDL No.: MFCD00002431

Formel: C₈H₈O₃
Molekylvægt: 152.15 g/mol
Kogepunkt: 280
Smeltepunkt: 99…103 °C
Massefylde: 1.38 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002431
CAS-nummer: 579-75-9
EINECS: 209-447-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR