4-Nitrobenzoesyre ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A14738.0B
A14738.0I
A14738.30
A14738.0BEA
885
DKK
A14738.0B
A14738.0I
A14738.30
4-Nitrobenzoesyre ≥99%
4-Nitrobenzoesyre
Purity: 99%
CAS No.: 62-23-7
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Melting Point: 239-241°
Beilstein Reference: 973593
Merck Reference: 14,6589
MDL No.: MFCD00007352
Formel:
C₇H₅NO₄ Kogepunkt: 359 °C (1013 hPa) Smeltepunkt: 239…241 °C Massefylde: 1,610 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007352 CAS-nummer: 62-23-7 EINECS: 200-526-2 Merck Index: 14,6589 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...