3,5-Diaminobenzoesyre 98% may contain up to 3% moisture
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 535-87-5
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Flash Point: 210°(410°F)
Melting Point: ca 235° dec.
Beilstein Reference: 2086484
Merck Reference: 14,2978
MDL No.: MFCD00007807
Formel:
C₇H₈N₂O₂ Kogepunkt: 546 °C (1013 hPa) Smeltepunkt: ∼235 °C (dec.) Massefylde: 1,45 g/cm³ (20 °C) Flammepunkt: 210 °C (410 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007807 CAS-nummer: 535-87-5 EINECS: 208-621-0 Merck Index: 14,2978 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...