4-Amino-2-hydroxybenzoesyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Amino-2-hydroxybenzoic acid

B23289.22 B23289.36
B23289.22EA 539 DKK
B23289.22 B23289.36
4-Amino-2-hydroxybenzoesyre ≥98%
4-Amino-2-hydroxybenzoesyre

Purity: 98+% CAS No.: 65-49-6 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Odour: Odourless Melting Point: ca 135-145° dec. Beilstein Reference: 473071 Merck Reference: 14,477 MDL No.: MFCD00007789

Formel: C₇H₇NO₃
Molekylvægt: 153.14 g/mol
Kogepunkt: 347
Smeltepunkt: ∼135…145 °C (dec.)
Massefylde: 1.545 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007789
CAS-nummer: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR