3-Aminosalicylsyre 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Aminosalicylic Acid, 3-Amino-2-hydroxybenzoic acid, 3-Amino-2-hydroxybenzoic acid
225262500
225260010
225260500
ACRO225262500EA
1190
DKK
ACRO225262500
ACRO225260010
ACRO225260500
3-Aminosalicylsyre 97%
3-Aminosalicylsyre
Formel:
C₇H₇NO₃ Molekylvægt: 153.14 g/mol Kogepunkt: 347 Smeltepunkt: 240 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010299 CAS-nummer: 570-23-0 EINECS: 209-328-0 |
Specifikationer Testresultater
Appearance | Beige to grey to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...