Natriumbenzoat 99+%, extra pure
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzoic acid sodium salt
148980010
148980050
ACRO148980010EA
372
DKK
ACRO148980010
ACRO148980050
Natriumbenzoat 99+%, extra pure
Natriumbenzoat
Formel:
C₆H₅COONa Smeltepunkt: >300 °C Massefylde: 1,44 g/cm³ (20 °C) Flammepunkt: ≥ 100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012463 CAS-nummer: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Specifikationer Testresultater
Appearance | White Crystalline powder or crystals or granules or flakes |
Infrared spectrum | Conforms |
Water | ≤2.0 % (K.F.) |
Acidity/alkalinity | ≤0.2 ml 0.1 M NaOH or HCl per gram |
Titration with HClO4 | ≥99.0 % (On dried substance) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...