Ostwald-viskometre
Leverandør: SI Analytics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
285404006 285404039 285404047 285404014 285404022
SCOI285404006EA 1960 DKK
SCOI285404006 SCOI285404039 SCOI285404047 SCOI285404014 SCOI285404022
Ostwald-viskometre
Viskosimetre Kapillærviskosimetere
Ostwald viscometers suitable for measurements of transparent liquids manual measurements, foaming liquids and liquid mixture with highly volatile components.