VWR® CalBox®, Calibration Weights, Class F1, Certified
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-2428EA 3880 DKK
611-2428 611-2429 611-2430 611-2431 611-2432 611-2433 611-2434 611-2435 611-2436 611-2437
VWR® CalBox®, Calibration Weights, Class F1, Certified
Lodder Kalibreringslodder
An economical range of calibration sets, with only three stainless steel weights, to meet routine calibration requirements of precision and analytical balances.
  • Individual serial number and calibration certificate
  • Each weight, from 1 g upwards, has a unique identification number for traceability
  • Laser engraving improves legibility of the markings
Leveringsinformation: Weights supplied in modern aluminium case with tweezers and balance brush.