Benzonitril ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Cyanobenzene, Phenylcyanide
A11173.0E
A11173.22
A11173.36
A11173.0EEA
2330
DKK
A11173.0E
A11173.22
A11173.36
Benzonitril ≥99%
Benzonitril
Purity: 99%
CAS No.: 100-47-0
Molecular Formula: C7H5N
Formula Weight: 103.12
Flash Point: 71°(159°F)
Refractive Index: 1.5285
Melting Point: -13°
Boiling Point: 191-193°
Beilstein Reference: 506893
Merck Reference: 14,1097
UN No.: 2224
MDL No.: MFCD00001770
Formel:
C₇H₅N Kogepunkt: 191…193 °C Smeltepunkt: –13 °C Massefylde: 1,007 Flammepunkt: 71 °C (159 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001770 CAS-nummer: 100-47-0 EINECS: 202-855-7 UN nummer: 2224 ADR: 6.1,II Merck Index: 14,1097 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...