Benzonitril 99%, pure
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Cyanobenzene, Phenylcyanide
105540010
105540025
ACRO105540010EA
582
DKK
ACRO105540010
ACRO105540025
Benzonitril 99%, pure
Benzonitril
Formel:
C₇H₅N Molekylvægt: 103.12 g/mol Kogepunkt: 188…191 °C (760 mmHg) Smeltepunkt: –13 °C Massefylde: 1.01 g/cm³ Flammepunkt: 71 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001770 CAS-nummer: 100-47-0 EINECS: 202-855-7 UN nummer: 2224 ADR: 6.1 II Merck Index: 13,01098 |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5270 to 1.5290 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with HClO4 | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...