4-Phenoxybenzaldehyd 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
198930050
198930250
ACRO198930050EA
607
DKK
ACRO198930050
ACRO198930250
4-Phenoxybenzaldehyd 98%
4-Phenoxybenzaldehyde
Formel:
C₁₃H₁₀O₂ Kogepunkt: 185 °C (14 mmHg) Smeltepunkt: 24…25 °C Massefylde: 1,132 g/cm³ Flammepunkt: >112 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003383 CAS-nummer: 67-36-7 EINECS: 200-650-7 |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.6100 to 1.6120 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...