VWR®, Funnels, with long stem
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
221-0172EA 270 DKK
221-0172 221-0173 221-0174
VWR®, Funnels, with long stem
Tragte Filtreringstragte Filtertragte
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Smooth internal walls.
Certificeringer: ISO 4798.