Multi-channel pipettes, Multipet
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1326.10 1328.10 1417.10 1418.10
KART1326.10EA 12570 DKK
KART1326.10 KART1328.10 KART1417.10 KART1418.10
Multi-channel pipettes, Multipet
Pipetter Automatiske pipetter
These multi-channel micropipettes with air cushion are ideal for dispensing in multiwell plates for ELISA dosages, molecular biology and DNA amplifications.
  • Volume can be easily selected by turning the emptying button or the cylinder
  • Large display ensures easy reading of the set volume value
  • Cones are indipendent, so the required force for securing the 8 or 12 tips is uniformly distributed
  • Minimum pressure required for inserting and ejecting tips
  • Movement of the cones allows the micropipette to oscillate during the loading operations
  • Lower part can freely turn 360°, the ejector is easily adjustable to house a large variety of tips and can be removed for cleaning
  • Fully autoclavable at +121 °C for 20 minutes, without having to recalibrate them
Certificeringer: EC/IVD-compliant, in accordance with EN IS 8655
Leveringsinformation: Supplied with individual calibration certificates.