VWR® Traceable®, Triple-Display Thermometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-1822EA 1430 DKK
620-1822 620-1823 620-1827
VWR® Traceable®, Triple-Display Thermometers
Termometre Digital termometre
These highly accurate thermometers monitor temperatures simultaneously in refrigerators and freezers, two refrigerator locations. Temperature-buffered bottle sensor eliminates transient temperature changes when refrigerator door is opened. Patented unit fulfils all CDC thermometer and vaccine thermometer requirements. They may also be used in water baths, heating blocks and incubators.
  • Displays exact time and date when an alarm is triggered, alarms are programmable in 0,1 °C increments
  • Monitor minimum and maximum readings and displays exact time and date